4-(2-Nitroanilino)phenol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4'-Hydroxy-2-nitrodiphenylamine, 4-[(2-Nitrophenyl)amino]phenol

N1185-1G N1185-5G
TCN1185-1GEA 26 USD
TCN1185-1G TCN1185-5G
4-(2-Nitroanilino)phenol ≥98.0% (by GC)
4-(2-Nitroanilino)phenol
Formula: C₁₂H₁₀N₂O₃
MW: 230.22 g/mol
MDL Number: MFCD00239470
CAS Number: 54381-08-7
EINECS: 259-132-4

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 145.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR