2-Acetyl-3,5-dimethylpyrazine ≥98.0% (by GC) (contains 2-Acetyl-3,6-dimethylpyrazine)

Supplier: TCI America
Warning

Synonyms: 1-(3,5-Dimethylpyrazin-2-yl)ethanone

A3326-5G A3326-1G
TCA3326-5GEA 121 USD
TCA3326-5G TCA3326-1G
2-Acetyl-3,5-dimethylpyrazine ≥98.0% (by GC) (contains 2-Acetyl-3,6-dimethylpyrazine)
2-Acetyl-3,5-dimethylpyrazine
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Flash Pt: 91°C
MDL Number: MFCD00055023
CAS Number: 54300-08-2
EINECS: 259-076-0

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 % (total of isomer)
Specific gravity (20/20) 1.0740 to 1.0840

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR