2-Iododibenzofuran ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Iododibenzo[b,d]furan

I0968-1G I0968-5G
TCI0968-1GEA 249 USD
TCI0968-1G TCI0968-5G
2-Iododibenzofuran ≥98.0% (by GC)
2-Iododibenzofuran
Formula: C₁₂H₇IO
MW: 294.09 g/mol
MDL Number: MFCD00092649
CAS Number: 5408-56-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR