2-Phenylethylamine hydrobromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Phenylethylammonium bromide, PEABr, Phenethylamine hydrobromide
P2388-1G
P2388-5G
TCP2388-1GEA
79
USD
TCP2388-1G
TCP2388-5G
2-Phenylethylamine hydrobromide ≥98.0% (by HPLC, titration analysis)
2-Phenylethylamine hydrobromide
Formula:
C₈H₁₁N·HBr MW: 202.10 g/mol Melting Pt: 259°C |
CAS Number:
53916-94-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...