2-Bromodibenzothiophene 5,5-Dioxide ≥95.0% (by GC)

Supplier: TCI America

B5267-1G B5267-200MG
TCB5267-1GEA 410 USD
TCB5267-1G TCB5267-200MG
2-Bromodibenzothiophene 5,5-Dioxide ≥95.0% (by GC)
2-Bromodibenzothiophene 5,5-Dioxide
Formula: C₁₂H₇BrO₂S
Melting Pt: 267°C
MDL Number: MFCD00623266
CAS Number: 53846-85-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR