2,6-Diphenylbenzo[1,2-b:4,5-b']difuran
Supplier: TCI America
D4633-1G
D4633-200MG
TCD4633-1GEA
708.2
USD
TCD4633-1G
TCD4633-200MG
2,6-Diphenylbenzo[1,2-b:4,5-b']difuran
2,6-Diphenylbenzo[1,2-b:4,5-b']difuran
Formula:
C₂₂H₁₄O₂ MW: 310.35 g/mol Melting Pt: 345°C |
CAS Number:
5379-77-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Elemental analysis (Carbon) | 82.50 to 87.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...