2-Methyldibenzo[f,h]quinoxaline ≥98.0% (by GC)
Supplier: TCI America
M3005-1G
M3005-200MG
TCM3005-1GEA
225
USD
TCM3005-1G
TCM3005-200MG
2-Methyldibenzo[f,h]quinoxaline ≥98.0% (by GC)
2-Methyldibenzo[f,h]quinoxaline
Formula:
C₁₇H₁₂N₂ MW: 244.30 g/mol |
MDL Number:
MFCD16038624 CAS Number: 536753-86-3 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...