4-(4-Bromophenyl)dibenzothiophene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-(4-Bromophenyl)dibenzo[b,d]thiophene

B5384-1G B5384-200MG
TCB5384-1GEA 183 USD
TCB5384-1G TCB5384-200MG
4-(4-Bromophenyl)dibenzothiophene ≥98.0% (by GC)
4-(4-Bromophenyl)dibenzothiophene
Formula: C₁₈H₁₁BrS
MW: 339.25 g/mol
MDL Number: MFCD19441307
CAS Number: 530402-77-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR