1-(2,6-Dimethylphenoxy)-2-propanone ≥95.0% (by GC)

Supplier: TCI America

Synonyms: (2,6-Dimethylphenoxy)acetone

D5121-1G D5121-5G
76219-164EA 48 USD
76219-164 76219-166
1-(2,6-Dimethylphenoxy)-2-propanone ≥95.0% (by GC)
1-(2,6-Dimethylphenoxy)-2-propanone
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 113°C
MDL Number: MFCD00130266
CAS Number: 53012-41-2
EINECS: 258-292-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR