4-Butoxy-4'-cyanobiphenyl ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4300-25G
B4300-5G
TCB4300-25GEA
448.8
USD
TCB4300-25G
TCB4300-5G
4-Butoxy-4'-cyanobiphenyl ≥98.0% (by GC)
4-Butoxy-4'-cyanobiphenyl
Formula:
C₁₇H₁₇NO |
MDL Number:
MFCD00183755 CAS Number: 52709-87-2 EINECS: 258-123-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Mesomorphic range | 75.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...