Carteolol hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone hydrochloride

C3015-250MG C3015-50MG
TCC3015-250MGEA 403 USD
TCC3015-250MG TCC3015-50MG
Carteolol hydrochloride ≥98.0% (by HPLC, titration analysis)
Carteolol hydrochloride
Formula: C₁₆H₂₄N₂O₃·HCl
MW: 328.84 g/mol
Melting Pt: 278°C
MDL Number: MFCD00941499
CAS Number: 51781-21-6
EINECS: 257-415-7
Merck Index: 14,01866

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR