9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
B4979-1G
B4979-200MG
TCB4979-1GEA
240
USD
TCB4979-1G
TCB4979-200MG
9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene ≥97.0% (by HPLC, total nitrogen)
9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene
Formula:
C₅₇H₄₀N₂ MW: 752.96 g/mol |
CAS Number:
510775-24-3 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...