Tiapride hydrochloride ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

T3600-1G T3600-250MG
TCT3600-1GEA 134 USD
TCT3600-1G TCT3600-250MG
Tiapride hydrochloride ≥99.0% (by HPLC, titration analysis)
Tiapride hydrochloride
Formula: C₁₅H₂₄N₂O₄S·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00133861
CAS Number: 51012-33-0
EINECS: 256-908-4
Merck Index: 14,09421

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR