3,4-Diacetyl-2,5-hexanedione ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Bis(acetylacetone), 1,1,2,2-Tetraacetylethane, 3,4-Diacetylhexane-2,5-dione

D4826-1G D4826-200MG
TCD4826-1GEA 597 USD
TCD4826-1G TCD4826-200MG
3,4-Diacetyl-2,5-hexanedione ≥98.0% (by GC)
3,4-Diacetyl-2,5-hexanedione
Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
Boiling Pt: 122°C
MDL Number: MFCD00031531
CAS Number: 5027-32-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 191.0 to 195.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR