1,4-Diazabicyclo[2.2.2]octane dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
|
Warning
|
D5251-1G
D5251-5G
76219-880EA
85
USD
76219-880
76219-882
1,4-Diazabicyclo[2.2.2]octane dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
1,4-Diazabicyclo[2.2.2]octane dihydrochloride
Formula:
C₆H₁₂N₂·₂HCl MW: 185.09 g/mol Melting Pt: 320°C |
CAS Number:
49563-87-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...