N,N'-Bis(3,5-dimethylphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥95.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Pigment Red 149

B4231-1G B4231-5G
TCB4231-1GEA 30 USD
TCB4231-1G TCB4231-5G
N,N'-Bis(3,5-dimethylphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥95.0% (by total nitrogen basis)
N,N'-Bis(3,5-dimethylphenyl)-3,4,9,10-perylenetetracarboxylic diimide
Formula: C₄₀H₂₆N₂O₄
MW: 598.66 g/mol
MDL Number: MFCD01318472
CAS Number: 4948-15-6
EINECS: 225-590-9

Order Now

Specification Test Results

Appearance Orange to Red powder to crystal
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR