2-Dodecylthiophene ≥97.0% (by GC)

Supplier: TCI America

D4874-1ML D4874-5ML
TCD4874-1MLEA 71 USD
TCD4874-1ML TCD4874-5ML
2-Dodecylthiophene ≥97.0% (by GC)
2-Dodecylthiophene
Formula: C₁₆H₂₈S
Boiling Pt: 155°C
MDL Number: MFCD22493346
CAS Number: 4861-61-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR