Tris(phenylthio)methane ≥98.0% (by GC)

Supplier: TCI America
Warning

T3126-25G T3126-5G
TCT3126-25GEA 348 USD
TCT3126-25G TCT3126-5G
Tris(phenylthio)methane ≥98.0% (by GC)
Tris(phenylthio)methane
Formula: C₁₉H₁₆S₃
MW: 340.52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010396
CAS Number: 4832-52-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR