Isoindolin-1-one ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,3-Dihydro-1H-isoindol-1-one, 1-Isoindolinone

I1064-1G I1064-5G
TCI1064-1GEA 82 USD
TCI1064-1G TCI1064-5G
Isoindolin-1-one ≥98.0% (by GC)
Isoindolin-1-one
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 337°C
MDL Number: MFCD03085939
CAS Number: 480-91-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 149.0 to 153.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR