1-Butyl-4-methylpyridinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

B5763-1G B5763-5G
TCB5763-1GEA 78 USD
TCB5763-1G TCB5763-5G
1-Butyl-4-methylpyridinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Butyl-4-methylpyridinium bis(trifluoromethanesulfonyl)imide
Formula: C₁₂H₁₆F₆N₂O₄S₂
MW: 430.38 g/mol
Melting Pt: 10°C
MDL Number: MFCD16038147
CAS Number: 475681-62-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR