2-Bromo-9,9-di-p-tolyl-9H-fluorene ≥98.0% (by GC)
Supplier: TCI America
B5619-1G
B5619-5G
TCB5619-1GEA
156
USD
TCB5619-1G
TCB5619-5G
2-Bromo-9,9-di-p-tolyl-9H-fluorene ≥98.0% (by GC)
2-Bromo-9,9-di-p-tolyl-9H-fluorene
Formula:
C₂₇H₂₁Br MW: 425.37 g/mol |
MDL Number:
MFCD16619315 CAS Number: 474918-33-7 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 182.0 to 186.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...