9-Bromo-10-(1,1':3',1''-terphenyl-5'-yl)anthracene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 9-([1,1':3',1''-Terphenyl]-5'-yl)-10-bromoanthracene

B4755-1G B4755-200MG
TCB4755-1GEA 224.9 USD
TCB4755-1G TCB4755-200MG
9-Bromo-10-(1,1':3',1''-terphenyl-5'-yl)anthracene ≥98.0% (by HPLC)
9-Bromo-10-(1,1':3',1''-terphenyl-5'-yl)anthracene
Formula: C₃₂H₂₁Br
MW: 485.42 g/mol
MDL Number: MFCD26406220
CAS Number: 474688-74-9

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR