10,10'-Dimethyl-9,9'-biacridinium bis(monomethyl Terephthalate) for chemiluminescence research

Supplier: TCI America
Warning

Synonyms: MMT

B4339-100MG B4339-1G
TCB4339-100MGEA 36 USD
TCB4339-100MG TCB4339-1G
10,10'-Dimethyl-9,9'-biacridinium bis(monomethyl Terephthalate) for chemiluminescence research
10,10'-Dimethyl-9,9'-biacridinium bis(monomethyl Terephthalate)
Formula: C₄₆H₃₆N₂O₈
MW: 744.80 g/mol
CAS Number: 469865-01-8

Order Now

Specification Test Results

Chemiluminescence test (Superoxide radical) to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR