4-Chloroquinoline 1-Oxide ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C3682-1G C3682-250MG
TCC3682-1GEA 252 USD
TCC3682-1G TCC3682-250MG
4-Chloroquinoline 1-Oxide ≥97.0% (by GC, titration analysis)
4-Chloroquinoline 1-Oxide
Formula: C₉H₆ClNO
MDL Number: MFCD18377747
CAS Number: 4637-59-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR