Dihydrolipoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D4586-1G D4586-200MG
TCD4586-1GEA 483 USD
TCD4586-1G TCD4586-200MG
Dihydrolipoic acid ≥97.0% (by GC, titration analysis)
Dihydrolipoic acid
Formula: C₈H₁₆O₂S₂
Boiling Pt: 175°C
Storage Temperature: Freezer
MDL Number: MFCD00166974
CAS Number: 462-20-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR