5-Azoniaspiro[4.4]nonane iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A3093-1G A3093-5G
TCA3093-1GEA 73 USD
TCA3093-1G TCA3093-5G
5-Azoniaspiro[4.4]nonane iodide ≥98.0% (by titrimetric analysis)
5-Azoniaspiro[4.4]nonane iodide
Formula: C₈H₁₆IN
CAS Number: 45650-35-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 95.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR