Aegeline ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Aegelin, N-(2-Hydroxy-2-(4-methoxyphenyl)ethyl)cinnamamide, (E)-N-[2-Hydroxy-2-(4-methoxyphenyl)ethyl]cinnamamide

A3170-25G A3170-5G
TCA3170-25GEA 266 USD
TCA3170-25G TCA3170-5G
Aegeline ≥98.0% (by HPLC, total nitrogen)
Aegeline
Formula: C₁₈H₁₉NO₃
MW: 297.35 g/mol
MDL Number: MFCD00048045
CAS Number: 456-12-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 177.0 to 181.0 deg-C
Specific rotation [a]20/D +49.0 to +53.0 deg (C=0.01, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR