2,7-Dibromo-9-phenylcarbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

D4832-1G D4832-200MG
TCD4832-1GEA 103 USD
TCD4832-1G TCD4832-200MG
2,7-Dibromo-9-phenylcarbazole ≥98.0% (by GC)
2,7-Dibromo-9-phenylcarbazole
Formula: C₁₈H₁₁Br₂N
MDL Number: MFCD28147705
CAS Number: 444796-09-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 179.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR