Formoterol Fumarate hydrate

Supplier: TCI America

F0881-10MG F0881-50MG
TCF0881-10MGEA 130 USD
TCF0881-10MG TCF0881-50MG
Formoterol Fumarate hydrate
Formoterol Fumarate hydrate
Formula: C₁₉H₂₄N₂O₄·½C₄H₄O₄·xH₂O
Storage Temperature: Freezer
MDL Number: MFCD03093863
CAS Number: 43229-80-7
Merck Index: 14,04245

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 % (total of 2peaks)
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water 1.5 to 5.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR