4,4-Diphenylcyclohexanol ≥98.0% (by GC)
Supplier: TCI America
D4973-1G
D4973-5G
TCD4973-1GEA
134
USD
TCD4973-1G
TCD4973-5G
4,4-Diphenylcyclohexanol ≥98.0% (by GC)
4,4-Diphenylcyclohexanol
Formula:
C₁₈H₂₀O |
MDL Number:
MFCD00087400 CAS Number: 42420-85-9 |
Specification Test Results
Purity (GC) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...