4-Fluoro-2-iodo-1-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

F1178-1G F1178-5G
TCF1178-1GEA 35 USD
TCF1178-1G TCF1178-5G
4-Fluoro-2-iodo-1-nitrobenzene ≥98.0% (by GC)
4-Fluoro-2-iodo-1-nitrobenzene
Formula: C₆H₃FINO₂
MW: 267.00 g/mol
Boiling Pt: 148°C
Storage Temperature: Refrigerator
MDL Number: MFCD16658618
CAS Number: 41860-64-4

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR