5-Chloro-2,3-diphenylpyrazine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C3205-1G
C3205-5G
TCC3205-1GEA
80
USD
TCC3205-1G
TCC3205-5G
5-Chloro-2,3-diphenylpyrazine ≥98.0% (by GC)
5-Chloro-2,3-diphenylpyrazine
Formula:
C₁₆H₁₁ClN₂ Boiling Pt: 145°C |
MDL Number:
MFCD00234892 CAS Number: 41270-66-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...