1,5-Diiodo-2,4-dimethylbenzene ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4,6-Diiodo-m-xylene

D5005-1G D5005-5G
76219-124EA 47 USD
76219-124 76219-126
1,5-Diiodo-2,4-dimethylbenzene ≥97.0% (by GC)
1,5-Diiodo-2,4-dimethylbenzene
Formula: C₈H₈I₂
MW: 357.96 g/mol
CAS Number: 4102-50-5

Order Now

Specification Test Results

Purity (GC) min. 97.0 %
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR