Thieno[3,2-b]thiophene-2-carbonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Cyanothieno[3,2-b]thiophene

T3221-1G T3221-200MG
TCT3221-1GEA 299 USD
TCT3221-1G TCT3221-200MG
Thieno[3,2-b]thiophene-2-carbonitrile ≥98.0% (by GC)
Thieno[3,2-b]thiophene-2-carbonitrile
Formula: C₇H₃NS₂
MW: 165.23 g/mol
MDL Number: MFCD08059461
CAS Number: 40985-58-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR