Methyl 4-Acetamido-2-methoxybenzoate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Acetamido-o-anisic acid methyl ester, 4-Acetamido-2-methoxybenzoic acid methyl ester, Methyl 4-Acetamido-o-anisate, Methyl 4-acetamido-2-methoxybenzoate
M2776-25G
M2776-5G
TCM2776-25GEA
87
USD
TCM2776-25G
TCM2776-5G
Methyl 4-Acetamido-2-methoxybenzoate ≥98.0% (by GC)
Methyl 4-Acetamido-2-methoxybenzoate
Formula:
C₁₁H₁₃NO₄ MW: 223.23 g/mol |
MDL Number:
MFCD00065258 CAS Number: 4093-29-2 EINECS: 223-839-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...