Spiro[fluorene-9,9'-xanthene]-3',6'-diol ≥98.0% (by GC)

Supplier: TCI America

S0953-1G S0953-5G
TCS0953-1GEA 162 USD
TCS0953-1G TCS0953-5G
Spiro[fluorene-9,9'-xanthene]-3',6'-diol ≥98.0% (by GC)
Spiro[fluorene-9,9'-xanthene]-3',6'-diol
Formula: C₂₅H₁₆O₃
CAS Number: 4081-00-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR