4-Bromo-2,6-diphenylpyrimidine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4943-1G B4943-200MG
TCB4943-1GEA 219.7 USD
TCB4943-1G TCB4943-200MG
4-Bromo-2,6-diphenylpyrimidine ≥98.0% (by GC)
4-Bromo-2,6-diphenylpyrimidine
Formula: C₁₆H₁₁BrN₂
MDL Number: MFCD00234885
CAS Number: 40734-24-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR