N,N'-Dicyclohexyl-1,2-ethanediamine hydrate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

D4518-25G D4518-5G
TCD4518-25GEA 603 USD
TCD4518-25G TCD4518-5G
N,N'-Dicyclohexyl-1,2-ethanediamine hydrate ≥98.0% (by GC, titration analysis)
N,N'-Dicyclohexyl-1,2-ethanediamine hydrate
Formula: C₁₄H₂₈N₂·xH₂O
Boiling Pt: 312°C
CAS Number: 4013-98-3
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 3.5 to 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR