2-(4-Bromophenyl)thiophene ≥97.0% (by GC)
Supplier: TCI America
B5106-1G
B5106-5G
76218-818EA
155
USD
76218-818
76218-820
2-(4-Bromophenyl)thiophene ≥97.0% (by GC)
2-(4-Bromophenyl)thiophene
Formula:
C₁₀H₇BrS MW: 239.13 g/mol |
MDL Number:
MFCD05664340 CAS Number: 40133-22-0 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 96.0 to 100.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...