5-Bromo-1,10-phenanthroline ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B5567-1G
B5567-200MG
TCB5567-1GEA
424
USD
TCB5567-1G
TCB5567-200MG
5-Bromo-1,10-phenanthroline ≥98.0% (by titrimetric analysis)
5-Bromo-1,10-phenanthroline
Formula:
C₁₂H₇BrN₂ Melting Pt: 119°C |
MDL Number:
MFCD00046903 CAS Number: 40000-20-2 EINECS: 254-742-7 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Water | max. 2.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...