2'-Hydroxy-4',6'-dimethoxy-2-phenylacetophenone ≥94.0% (by GC)

Supplier: TCI America

H1577-1G H1577-5G
TCH1577-1GEA 165 USD
TCH1577-1G TCH1577-5G
2'-Hydroxy-4',6'-dimethoxy-2-phenylacetophenone ≥94.0% (by GC)
2'-Hydroxy-4',6'-dimethoxy-2-phenylacetophenone
Formula: C₁₆H₁₆O₄
MDL Number: MFCD00218596
CAS Number: 39604-66-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 94.0 %
Melting point 114.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR