Carboxymethyldextran sodium salt, M.W. ~ 10000
Supplier: TCI America
C3250-1G
C3250-5G
TCC3250-1GEA
42
USD
TCC3250-1G
TCC3250-5G
Carboxymethyldextran sodium salt, M.W. ~ 10000
Carboxymethyldextran sodium salt
MDL Number:
MFCD00146477 CAS Number: 39422-83-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Content (Sodium) | 6.5 to 9.5 % (calcd.on dried substance) |
Drying loss | max. 8.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...