N'-(4-Bromo-2-cyanophenyl)-N,N-dimethylformamidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4362-1G
B4362-5G
TCB4362-1GEA
155.9
USD
TCB4362-1G
TCB4362-5G
N'-(4-Bromo-2-cyanophenyl)-N,N-dimethylformamidine ≥98.0% (by GC, titration analysis)
N'-(4-Bromo-2-cyanophenyl)-N,N-dimethylformamidine
Formula:
C₁₀H₁₀BrN₃ |
CAS Number:
39255-60-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...