Bis(2-bromophenyl)acetylene ≥96.0% (by GC)
Supplier: TCI America
B4989-1G
B4989-5G
TCB4989-1GEA
203
USD
TCB4989-1G
TCB4989-5G
Bis(2-bromophenyl)acetylene ≥96.0% (by GC)
Bis(2-bromophenyl)acetylene
Formula:
C₁₄H₈Br₂ |
MDL Number:
MFCD00238712 CAS Number: 38399-13-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...