Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde ≥95.0% (by GC)

Supplier: TCI America

B5279-1G B5279-200MG
TCB5279-1GEA 334 USD
TCB5279-1G TCB5279-200MG
Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde ≥95.0% (by GC)
Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 73°C
Melting Pt: 13°C
Density: 1.04
Storage Temperature: Refrigerator
CAS Number: 38259-00-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 % (total of isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR