1-Methyl-1-pentylpyrrolidinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 1-Amyl-1-methylpyrrolidinium bis(trifluoromethanesulphonyl)imide

M3118-1G M3118-5G
TCM3118-1GEA 61 USD
TCM3118-1G TCM3118-5G
1-Methyl-1-pentylpyrrolidinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
1-Methyl-1-pentylpyrrolidinium bis(trifluoromethanesulfonyl)imide
Formula: C₁₂H₂₂F₆N₂O₄S₂
MW: 436.43 g/mol
Melting Pt: 8°C
CAS Number: 380497-17-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR