1-(2-Hydroxyethyl)-3-methylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

H1492-25G H1492-5G
TCH1492-25GEA 240.6 USD
TCH1492-25G TCH1492-5G
1-(2-Hydroxyethyl)-3-methylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1-(2-Hydroxyethyl)-3-methylimidazolium tetrafluoroborate
Formula: C₆H₁₁BF₄N₂O
MDL Number: MFCD12761438
CAS Number: 374564-83-7
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR