Oleyl Phosphate (Mono- and Di- ester mixture)
Supplier: TCI America
|
Warning
|
Synonyms:
Phosphoric acid Oleyl ester
O0146-25ML
O0146-500ML
TCO0146-25MLEA
31.53
USD
TCO0146-25ML
TCO0146-500ML
Oleyl Phosphate (Mono- and Di- ester mixture)
Oleyl Phosphate (Mono- and Di- ester mixture)
MDL Number:
MFCD00135103 CAS Number: 37310-83-1 EINECS: 253-455-4 |
Specification Test Results
Appearance | White or Colorless to Brown powder to lump to clear liquid |
Assay of mono ester | 37.0 to 50.0 % |
Assay of Diester | 50.0 to 63.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...