1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

T3457-25G T3457-5G
76220-654EA 279 USD
76220-654 76220-656
1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate ≥97.0% (by HPLC, total nitrogen)
1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate
Formula: C₁₆H₁₈F₆NP
MW: 369.29 g/mol
MDL Number: MFCD16621101
CAS Number: 372081-65-7

Order Now

Specification Test Results

Appearance White to Purple to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR