4-Methyl-N-tosylbenzenesulfonamide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Bis(p-tolylsulphonyl)amine, Ditosylamine, Bis(p-toluenesulphonyl)amine, Bis(4-methylbenzenesulphonyl)amine

M2975-1G M2975-5G
TCM2975-1GEA 41.7 USD
TCM2975-1G TCM2975-5G
4-Methyl-N-tosylbenzenesulfonamide ≥98.0% (by HPLC)
4-Methyl-N-tosylbenzenesulfonamide
Formula: C₁₄H₁₅NO₄S₂
MDL Number: MFCD00014917
CAS Number: 3695-00-9
EINECS: 223-016-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 171.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR